4-(Trifluoromethyl)benzeneboronic acid, 98%
4-(Trifluoromethyl)benzeneboronic acid, 98%
4-(Trifluoromethyl)benzeneboronic acid, 98%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzeneboronic acid, 98%

CAS: 128796-39-4 | C7H6BF3O2 | 189.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22374.03
also known as B22374-03
Price (USD)
70.65
Online Exclusive
78.80
Save 8.15 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.80
Save 8.15 (10%)
Each
Chemical Identifiers
CAS128796-39-4
IUPAC Name[4-(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC7H6BF3O2
InChI KeyALMFIOZYDASRRC-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥97.5%
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
View more
4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.

Solubility
Soluble in DMSO, not in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Cao KS.; Bian HX.; Zheng WH. Mild arylboronic acid catalyzed selective [4 + 3] cycloadditions: access to cyclohepta[b]benzofurans and cyclohepta[b]indoles. Org Biomol Chem. 2015, 13, (23), 6449-52.
  2. Maki T.; Ishihara K.; Yamamoto H. 4,5,6,7-Tetrachlorobenzo[d][1,3,2]dioxaborol- 2-ol as an effective catalyst for the amide condensation of sterically demanding carboxylic acids. Org Lett.2006, 8, (7), 1431-4.
  3. For phosphine-free palladium-catalyzed Suzuki coupling with aryl halides, see: J. Org. Chem., 59, 5034 (1994).