2-(Diphenylphosphino)benzoic acid, 97%
2-(Diphenylphosphino)benzoic acid, 97%
2-(Diphenylphosphino)benzoic acid, 97%
Thermo Scientific Chemicals

2-(Diphenylphosphino)benzoic acid, 97%

CAS: 17261-28-8 | C19H15O2P | 306.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22389.14
also known as B22389-14
Price (USD)
819.65
Online Exclusive
910.00
Save 90.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
819.65
Online Exclusive
910.00
Save 90.35 (10%)
Each
Chemical Identifiers
CAS17261-28-8
IUPAC Name2-(diphenylphosphanyl)benzoic acid
Molecular FormulaC19H15O2P
InChI KeyUYRPRYSDOVYCOU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Melting Point (clear melt)172.0-181.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only