S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%
S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%
S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%
Thermo Scientific Chemicals

S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%

CAS: 41840-28-2 | C11H16N2O2S | 240.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22435.06
also known as B22435-06
Price (USD)
23.70
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
23.70
Each
Chemical Identifiers
CAS41840-28-2
IUPAC Nametert-butyl [(4,6-dimethylpyrimidin-2-yl)sulfanyl]formate
Molecular FormulaC11H16N2O2S
InChI KeyPOTDIELOEHTPJN-UHFFFAOYSA-N
SMILESCC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to pale brown
FormCrystalline powder
Elemental AnalysisCarbon C: 52-78-57.17% (Theoretical 54.98%)
Elemental AnalysisHydrogen H: 6.44-6.98% (Theoretical 6.71%)
Elemental AnalysisSulfur S: 12.81-13.88% (Theoretical 13.34%)
View more
S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only
Reagent for protection of amino acids as their Boc derivatives in high yield under mild conditions: Bull. Chem. Soc. Jpn., 46, 1269 (1973). See Appendix 6.