2-Phenylpropionaldehyde, 97%
2-Phenylpropionaldehyde, 97%
2-Phenylpropionaldehyde, 97%
Thermo Scientific Chemicals

2-Phenylpropionaldehyde, 97%

CAS: 93-53-8 | C9H10O | 134.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B22436.22
also known as B22436-22
Price (USD)
53.50
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
53.50
Each
Chemical Identifiers
CAS93-53-8
IUPAC Name2-phenylpropanal
Molecular FormulaC9H10O
InChI KeyIQVAERDLDAZARL-UHFFFAOYNA-N
SMILESCC(C=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Free acid (titration)≤2.0%
Identification (FTIR)Conforms
Refractive Index1.5140-1.5240 @ 20?C
View more
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.

Solubility
Soluble in most fixed oils, propylene glycol. Insoluble in glycerin.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Richard F. Heck. The palladium-catalyzed arylation of enol esters, ethers, and halides. A new synthesis of 2-aryl aldehydes and ketones. J. Am. Chem. Soc. 1968, 90, (20), 5535-5538.
  2. Charles D. Thompson.; Michael T. Kinter.; Timothy L. Macdonald. Synthesis and in Vitro Reactivity of 3-Carbamoyl-2-phenylpropionaldehyde and 2-Phenylpropenal: Putative Reactive Metabolites of Felbamate. Chem. Res. Toxicol.1996, 9, (8), 1225-1229.