1-Hexylboronic acid, 97%
1-Hexylboronic acid, 97%
1-Hexylboronic acid, 97%
Thermo Scientific Chemicals

1-Hexylboronic acid, 97%

CAS: 16343-08-1 | C6H15BO2 | 129.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Catalog number B22446.88
also known as B22446-88
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.80
Save 12.15 (14%)
Each
Quantity:
2.5 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.80
Save 12.15 (14%)
Each
Chemical Identifiers
CAS16343-08-1
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms to structure
Proton NMRConforms to structure
Appearance (Color)White to pale cream
FormCrystalline powder
Assay (Aqueous acid-base Titration)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only