4-(Methylthio)benzyl alcohol, 98%
4-(Methylthio)benzyl alcohol, 98%
4-(Methylthio)benzyl alcohol, 98%
4-(Methylthio)benzyl alcohol, 98%
Thermo Scientific Chemicals

4-(Methylthio)benzyl alcohol, 98%

CAS: 3446-90-0 | C8H10OS | 154.227 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22455.14
also known as B22455-14
Price (USD)
141.65
Special offer
Online exclusive
Ends: 30-Jun-2026
167.00
Save 25.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
141.65
Special offer
Online exclusive
Ends: 30-Jun-2026
167.00
Save 25.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS3446-90-0
IUPAC Name[4-(methylsulfanyl)phenyl]methanol
Molecular FormulaC8H10OS
InChI KeyMTXQKSQYMREAGJ-UHFFFAOYSA-N
SMILESCSC1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)38.5-44.5?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Kandasamy Jeyakumar.; Dillip Kumar Chand. Selective oxidation of sulfides to sulfoxides and sulfones at room temperature using H2O2 and a Mo(VI) salt as catalyst. Tetrahedron Letters. 2006, 47, (27), 4573-4576.
  2. Won-Hee Kim.; In Soo Park.; Jaiwook Park. Acceptor-Free Alcohol Dehydrogenation by Recyclable Ruthenium Catalyst. Org. Lett. 2006, 8, (12), 2543-2545.