o-Thiocresol, 98%
o-Thiocresol, 98%
o-Thiocresol, 98%
Thermo Scientific Chemicals

o-Thiocresol, 98%

CAS: 137-06-4 | C7H8S | 124.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22460.22
also known as B22460-22
Price (USD)
402.65
Online Exclusive
447.00
Save 44.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
402.65
Online Exclusive
447.00
Save 44.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS137-06-4
IUPAC Name2-methylbenzene-1-thiol
Molecular FormulaC7H8S
InChI KeyLXUNZSDDXMPKLP-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5790-1.5830 @ 20?C
o-Thiocresol is used as a thiol for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Thiocresol is used as a thiol for proteomics research.

Solubility
Soluble in ether and ethanol. Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. E. Campaigne.; S. Osborn. Notes - Improved Procedure for the Preparation of Aromatic Thiols. J. Org. Chem. 1957, 22, (5), 561-562.
  2. Yasser M. Riyad.; Ralf Hermann.; Ortwin Brede. GarnierHidden triplet states of aromatic thiols characterized by radiation-chemical sensitizations. Chemical Physics Letters. 2003, 379, (5-6), 776-780.