6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%
Thermo Scientific Chemicals

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%

CAS: 2328-12-3 | C11H16ClNO2 | 229.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22469.14
also known as B22469-14
Price (USD)
113.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
113.00
Each
Add to cart
Chemical Identifiers
CAS2328-12-3
IUPAC Name6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Molecular FormulaC11H16ClNO2
InChI KeySHOWAGCIRTUYNA-UHFFFAOYSA-N
SMILES[Cl-].COC1=CC2=C(C[NH2+]CC2)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)249.0-258.0?C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only