1-Bromoperfluorooctane, 98+%
1-Bromoperfluorooctane, 98+%
1-Bromoperfluorooctane, 98+%
Thermo Scientific Chemicals

1-Bromoperfluorooctane, 98+%

CAS: 423-55-2 | C8BrF17 | 498.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22470.14
also known as B22470-14
Price (USD)
221.65
Online Exclusive
246.00
Save 24.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
221.65
Online Exclusive
246.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS423-55-2
IUPAC Name1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
Molecular FormulaC8BrF17
InChI KeyWTWWXOGTJWMJHI-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.3030-1.3070 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromoperfluorooctane 1-Bromoheptadecafluorooctane (perfluorooctyl bromide) can be used as contrast agent in the vascular system for fluorine-19 magnetic resonance imaging or as synthetic oxygen carrier. It is used as an O2 carrier in some emulsions.

Solubility
Not miscible or difficult to mix in water.

Notes
Incompatible with oxidizing agents, acids, metals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. H K Lee; O Nalcioglu. Reblurred deconvolution method for chemical shift removal in F-19 (PFOB) MR imaging. Journal of Magnetic Resonance Imaging. 1992, 2 (1), 53-61.
  2. Odile Diou; Elias Fattal; Vianney Delplace; Nicolas Mackiewicz; Julien Nicolas; Sébastien Mériaux; Julien Valette; Caroline Robic; Nicolas Tsapis. RGD decoration of PEGylated polyester nanocapsules of perfluorooctyl bromide for tumor imaging: Influence of pre or post-functionalization on capsule morphology. European Journal of Pharmaceutics and Biopharmaceutics. 2014, 87 (1), 170-177.