Methyl 2,5-dichlorobenzoate, 99%
Methyl 2,5-dichlorobenzoate, 99%
Methyl 2,5-dichlorobenzoate, 99%
Methyl 2,5-dichlorobenzoate, 99%
Thermo Scientific Chemicals

Methyl 2,5-dichlorobenzoate, 99%

CAS: 2905-69-3 | C8H6Cl2O2 | 205.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B22489.18
also known as B22489-18
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2905-69-3
IUPAC Namemethyl 2,5-dichlorobenzoate
Molecular FormulaC8H6Cl2O2
InChI KeySPJQBGGHUDNAIC-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystalline powder or lumpy powder or fused solid
Assay (GC)≥98.5%
Melting Point (clear melt)37-42?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only