Magnesium silicate, activated, for chromatography, 60-100 mesh
Magnesium silicate, activated, for chromatography, 60-100 mesh
Magnesium silicate, activated, for chromatography, 60-100 mesh
Thermo Scientific Chemicals

Magnesium silicate, activated, for chromatography, 60-100 mesh

CAS: 1343-88-0 | MgO3Si | 100.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B22490.0B
also known as B22490-0B
Price (USD)
327.65
Special offer
Online exclusive
Ends: 28-Dec-2026
385.00
Save 57.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
327.65
Special offer
Online exclusive
Ends: 28-Dec-2026
385.00
Save 57.35
Each
Chemical Identifiers
CAS1343-88-0
IUPAC Namesilicon(4+) magnesium(2+) trioxidandiide
Molecular FormulaMgO3Si
InChI KeyZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol)100.39
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Particle Size60-100 mesh (0.140-0.250mm): min. 90%
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with strong acids and hydrogen fluoride.
RUO – Research Use Only

General References:

  1. Hassanien, M. F. R.; Dohim, M. A.; Abdel-Salam, A. S.; Sitohhy, M. Z. Physicochemical characteristics and oxidative stability of de-acidified lampante high acid olive oil. J. Food Meas. Charact. 2015, 9 (4), 564-574.
  2. Pal, J.; Ghorai, S.; Goswami, M. C.; Prakash, S.; Venugopalan, T. Development of Pellet-Sinter Composite Agglomerate for Blast Furnace. ISIJ Int. 2014, 54 (3), 620-627.