(+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%
(+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%
(+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%
Thermo Scientific Chemicals

(+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%

CAS: 88568-95-0 | C13H18NO7P | 331.261 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22506.14
also known as B22506-14
Price (USD)
270.65
Online Exclusive
300.00
Save 29.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
270.65
Online Exclusive
300.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS88568-95-0
IUPAC Namemethyl (2S)-2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Molecular FormulaC13H18NO7P
InChI KeyGSYSFVSGPABNNL-NSHDSACASA-N
SMILESCOC(=O)[C@@H](NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)(by Carbon EA) : >96.0 to<104.0%
Elemental Analysis(Carbon) : 45.25-49.01% (Theory 47.14%)
Elemental Analysis(Hydrogen) : 5.25-5.69% (Theory 5.48%)
Proton NMRConforms to structure
Phosphorus-31 NMRConforms to structure
View more
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only