Zinc bromide, anhydrous, 98%
Zinc bromide, anhydrous, 98%
Zinc bromide, anhydrous, 98%
Thermo Scientific Chemicals

Zinc bromide, anhydrous, 98%

Zinc bromide, 98%, Br2Zn, CAS Number-7699-45-8, acmc-209p7h, zinc bromide, anhydrous 100g, zincbromide, zinc bromide hydrate, puratronic, ksc378m6n, zinc 2+ ion dibromide, zinc ii bromide, 500g, EM>9,, 650 deg.C, 4.201 | CAS: 7699-45-8 | Br2Zn | 225.188 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B22510.36
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
146.00
Save 21.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
146.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS7699-45-8
IUPAC Namezinc(2+) dibromide
Molecular FormulaBr2Zn
InChI KeyVNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES[Zn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥97.5% to ≤102.5%
Water Content (Karl Fischer Titration)≤1.0%
Zinc bromide solution can be used as a transparent shield against radiation. It is used as an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. It finds an application as Lewis acid in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc bromide solution can be used as a transparent shield against radiation. It is used as an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. It finds an application as Lewis acid in organic reactions.

Solubility
Soluble in alcohol, ether, acetone and tetrahydrofuran.

Notes
Hygroscopic.
RUO – Research Use Only

General References:

  1. Odabachian, Y.; Wang , Q.; Zhu, J. Tertiary Amines as Synthetic Equivalents of Vinyl Cations: Zinc Bromide Promoted Coupling of Propargylamines with alpha-Isocyanoacetamides To Give 2,4,5-Trisubstituted Oxazoles Initiated by an Internal Redox Process. Chem. Eur. J. 2013, 19 (37), 12229-12233.
  2. Tan, L.; Zhou, P.; Chen, C.; Liu, W. An efficient method for the construction of polysubstituted 4-pyridones via self-condensation of beta-keto amides mediated by P2O5 and catalyzed by zinc bromide. Beilstein J. Org. Chem. 2013, 9, 2681-2687.