2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B22515.22
also known as B22515-22
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS6976-37-0
IUPAC Name2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC8H19NO5
InChI KeyOWMVSZAMULFTJU-UHFFFAOYSA-N
SMILESOCCN(CCO)C(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5% to ≤102.5%
Identification (FTIR)Conforms
Appearance (Color)White to pale cream

It is used as a biological buffer, organic synthesize, chemical & pharmaceutical intermediate. It is suitable for fractionation of hemoglobin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a biological buffer, organic synthesize, chemical & pharmaceutical intermediate. It is suitable for fractionation of hemoglobin.

Solubility
Soluble in water (209.2 g/L), at 20°C and alcohol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, air.
RUO – Research Use Only

General References:

  1. K H Scheller; T H Abel; P E Polanyi; P K Wenk; B E Fischer; H Sigel. Metal ion/buffer interactions. Stability of binary and ternary complexes containing 2-[bis(2-hydroxyethyl)amino]-2(hydroxymethyl)-1,3-propanediol (Bistris) and adenosine 5'-triphosphate (ATP). European Journal of Biochemistry. 1980, 107 (2), 455-466.
  2. H Kosaka; I Tyuma. Production of superoxide anion by N,N-bis(2-hydroxyethyl)-iminotris(hydroxymethyl)methane buffer during oxidation of oxyhemoglobin by nitrite and effect of inositol hexaphosphate on the oxidation. Biochimica et Biophysica Acta. 1982, 709 (2), 187-193.
  3. Buffer: pH range 5.5 - 8.0.