Methyl 4-hydroxyphenylacetate, 98+%
Methyl 4-hydroxyphenylacetate, 98+%
Methyl 4-hydroxyphenylacetate, 98+%
Methyl 4-hydroxyphenylacetate, 98+%
Thermo Scientific Chemicals

Methyl 4-hydroxyphenylacetate, 98+%

CAS: 14199-15-6 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B22533.22
also known as B22533-22
Price (USD)
131.65
Online Exclusive
146.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
131.65
Online Exclusive
146.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS14199-15-6
IUPAC Namemethyl 2-(4-hydroxyphenyl)acetate
Molecular FormulaC9H10O3
InChI KeyXGDZEDRBLVIUMX-UHFFFAOYSA-N
SMILESCOC(=O)CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)54-59?C
Appearance (Color)White to cream or pale yellow
FormCrystals or crystalline powder
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only