Benzyl phenyl ether, 97%
Benzyl phenyl ether, 97%
Benzyl phenyl ether, 97%
Benzyl phenyl ether, 97%
Thermo Scientific Chemicals

Benzyl phenyl ether, 97%

CAS: 946-80-5 | C13H12O | 184.238 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22539.22
also known as B22539-22
Price (USD)
243.65
Special offer
Online exclusive
Ends: 30-Jun-2026
287.00
Save 43.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
243.65
Special offer
Online exclusive
Ends: 30-Jun-2026
287.00
Save 43.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS946-80-5
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to pale cream or pink
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)34.0-44.0?C
Benzyl phenyl ether reacts with aluminum bromide in chlorobenzene solution to afford a mixture of phenol, o-benzyl phenol and dichlorodiphenylmethane. It is a useful as model compound in catalytic chemistry to represent the a-O4 ether bond in lignin and coal. It contains a weak ether bond of 234kJ/mol and belongs to the most thermo-labile compounds in lignin and low rank coal. Influence of alkali carbonates, common additives in biomass conversion, on the reaction pathways of BPE in superheated water has been reported. Cesium-exchanged heteropolyacid catalyzed decomposition of benzyl phenyl ether to aromatics has been investigated. Photo-Claisen rearrangements of benzyl phenyl ether was investigated in cation-exchanged Y zeolites and polyethylenes of differing crystallinities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl phenyl ether reacts with aluminum bromide in chlorobenzene solution to afford a mixture of phenol, o-benzyl phenol and dichlorodiphenylmethane. It is a useful as model compound in catalytic chemistry to represent the a-O4 ether bond in lignin and coal. It contains a weak ether bond of 234kJ/mol and belongs to the most thermo-labile compounds in lignin and low rank coal. Influence of alkali carbonates, common additives in biomass conversion, on the reaction pathways of BPE in superheated water has been reported. Cesium-exchanged heteropolyacid catalyzed decomposition of benzyl phenyl ether to aromatics has been investigated. Photo-Claisen rearrangements of benzyl phenyl ether was investigated in cation-exchanged Y zeolites and polyethylenes of differing crystallinities.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Richard H.Schlosberg; William H.Davis Jr.; Terrence R.Ashe. Pyrolysis studies of organic oxygenates. 2. Benzyl phenyl ether pyrolysis under batch autoclave conditions. Fuel. 1981, 60,(3), 201-204.
  2. Virginia Roberts; Sebastian Fendt; Angeliki A.Lemonidou; Xuebing Li; Johannes A.Lercher. Influence of alkali carbonates on benzyl phenyl ether cleavage pathways in superheated water. Applied Catalysis B: Environmental. 2010, 95,(1-2), 71-77.