Di-n-decyl sulfide, 97%
Di-n-decyl sulfide, 97%
Di-n-decyl sulfide, 97%
Thermo Scientific Chemicals

Di-n-decyl sulfide, 97%

CAS: 693-83-4 | C20H42S | 314.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B22560.18
also known as B22560-18
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS693-83-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)22.0-32.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only