Flavanone, 98+%
Flavanone, 98+%
Flavanone, 98+%
Thermo Scientific Chemicals

Flavanone, 98+%

CAS: 487-26-3 | C15H12O2 | 224.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B22587.09
also known as B22587-09
Price (USD)
37.65
Online Exclusive
42.10
Save 4.45 (11%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
42.10
Save 4.45 (11%)
Each
Add to cart
Chemical Identifiers
CAS487-26-3
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Melting Point (clear melt)74.0-80.0?C
Appearance (Color)White
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only