2-Fluoro-alpha-methyl-4-biphenylacetic acid, 99%
2-Fluoro-alpha-methyl-4-biphenylacetic acid, 99%
2-Fluoro-alpha-methyl-4-biphenylacetic acid, 99%
Thermo Scientific Chemicals

2-Fluoro-alpha-methyl-4-biphenylacetic acid, 99%

CAS: 5104-49-4 | C15H13FO2 | 244.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22603.06
also known as B22603-06
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS5104-49-4
IUPAC Name2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Molecular FormulaC15H13FO2
InChI KeySYTBZMRGLBWNTM-UHFFFAOYNA-N
SMILESCC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)111.5-115.5?C
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White
FormPowder
A cyclooxygenase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only