Thiophene-2-thiol, contains dimer, 97% as monomer
Thiophene-2-thiol, contains dimer, 97% as monomer
Thiophene-2-thiol, contains dimer, 97% as monomer
Thiophene-2-thiol, contains dimer, 97% as monomer
Thermo Scientific Chemicals

Thiophene-2-thiol, contains dimer, 97% as monomer

CAS: 7774-74-5 | C4H4S2 | 116.196 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22642.06
also known as B22642-06
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS7774-74-5
IUPAC Namethiophene-2-thiol
Molecular FormulaC4H4S2
InChI KeySWEDAZLCYJDAGW-UHFFFAOYSA-N
SMILESSC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0% (as monomer)
CommentForms dimer (4-(2-thienylthio)tetrahydrothiophene-2-thione) on storage
CommentReforms monomer on distillation
CommentRI for low dimer content 1.6130-1.6210 @ 20?C
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only