Hydroquinone-O,O'-diacetic acid, 98%
Hydroquinone-O,O'-diacetic acid, 98%
Hydroquinone-O,O'-diacetic acid, 98%
Thermo Scientific Chemicals

Hydroquinone-O,O'-diacetic acid, 98%

CAS: 2245-53-6 | C10H10O6 | 226.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22645.06
also known as B22645-06
Price (USD)
47.65
Online exclusive
53.30
Save 5.65
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
47.65
Online exclusive
53.30
Save 5.65
Each
Chemical Identifiers
CAS2245-53-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Hydroquinone-O,O'-diacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone-O,O′-diacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Richard T.Pon; Shuyuan Yu. Hydroquinone-O,O'-diacetic acid (Q-linker) as a replacement for succinyl and oxalyl linker arms in solid phase oligonucleotide synthesis. Nucl. Acids Res. 1997, 25,(18), 3629-3635. 
  2. R. T. Pon; S.Yu; Z.Guo; X.Yang; Y.S.Sanghvi. Reusable solid-phase supports for oligonucleotide synthesis using hydroquinone-O,O'-diacetic acid (Q-linker). Nucleosides and Nucleotides. 1999, 18,(6-7), 1237-1238.
  3. Linker for solid-phase oligonucleotide synthesis: cleavage from the support occurs ca 30x more rapidly than with the conventional succinic acid linker: Nucleic Acids Res., 25, 3629; Bioconjugate Chem., 10, 1051 (1999).