BODIPY&trade; FL C<sub>5</sub>-Ceramide complexed to BSA
Invitrogen™
BODIPY™ FL C5-Ceramide complexed to BSA
BODIPY FL C5 ceramide complexed to BSA can be used in sphingolipid transport and metabolism mechanisms. Complexing fluorescent lipids withRead more
Have Questions?
Catalog NumberQuantity
B226505 mg
Catalog number B22650
Price (USD)
373.10
Special offer
Online exclusive
Ends: 31-Dec-2026
455.00
Save 81.90
Each
Quantity:
5 mg
Price (USD)
373.10
Special offer
Online exclusive
Ends: 31-Dec-2026
455.00
Save 81.90
Each
BODIPY FL C5 ceramide complexed to BSA can be used in sphingolipid transport and metabolism mechanisms. Complexing fluorescent lipids with bovine serum albuin (BSA) facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe - the BSA-complexed probe can be directly dissolved in water.
Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialSphingolipids
Recommended StorageStore in freezer (-5 to -30°C) and protect from light.
Physical FormSolid
Product LineBODIPY
Quantity5 mg
Unit SizeEach
Citations & References (130)
Citations & References
Abstract
Centrosome localization determines neuronal polarity.
Authors:de Anda FC, Pollarolo G, Da Silva JS, Camoletto PG, Feiguin F, Dotti CG
Journal:Nature
PubMed ID:16079847
'Neuronal polarization occurs shortly after mitosis. In neurons differentiating in vitro, axon formation follows the segregation of growth-promoting activities to only one of the multiple neurites that form after mitosis. It is unresolved whether such spatial restriction makes use of an intrinsic program, like during C. elegans embryo polarization, or ... More
Time-lapse analysis and mathematical characterization elucidate novel mechanisms underlying muscle morphogenesis.
Authors:Snow CJ, Goody M, Kelly MW, Oster EC, Jones R, Khalil A, Henry CA,
Journal:PLoS Genet
PubMed ID:18833302
'Skeletal muscle morphogenesis transforms short muscle precursor cells into long, multinucleate myotubes that anchor to tendons via the myotendinous junction (MTJ). In vertebrates, a great deal is known about muscle specification as well as how somitic cells, as a cohort, generate the early myotome. However, the cellular mechanisms that generate ... More
Two-photon fluorescence absorption and emission spectra of dyes relevant for cell imaging.
Authors:Bestvater F, Spiess E, Stobrawa G, Hacker M, Feurer T, Porwol T, Berchner-Pfannschmidt U, Wotzlaw C, Acker H
Journal:J Microsc
PubMed ID:12423261
'Two-photon absorption and emission spectra for fluorophores relevant in cell imaging were measured using a 45 fs Ti:sapphire laser, a continuously tuneable optical parametric amplifier for the excitation range 580-1150 nm and an optical multichannel analyser. The measurements included DNA stains, fluorescent dyes coupled to antibodies as well as organelle ... More
Use of fluorescent dyes for measurement and localization of organelles associated with Ca2+ store release in human neutrophils.
Authors:Davies EV, Blanchfield H, Hallett MB
Journal:Cell Biol Int
PubMed ID:9693835
'Fura-2 and its lipid analogue, FFP-18, were used to measure changes in cytosolic free Ca2+ concentration within human neutrophils. Whereas fura-2 was employed to monitor cytosolic Ca2+ increases throughout the cytosol, FFP-18 was used to monitor Ca2+ changes only near the membrane. This latter probe was incorporated into the plasma ... More
Changes in architecture of the Golgi complex and other subcellular organelles during myogenesis.
Authors:Ralston E
Journal:J Cell Biol
PubMed ID:7678420
'Myogenesis involves changes in both gene expression and cellular architecture. Little is known of the organization, in muscle in vivo, of the subcellular organelles involved in protein synthesis despite the potential importance of targeted protein synthesis for formation and maintenance of functional domains such as the neuromuscular junction. A panel ... More