4-Hydroxy-2,2,6,6-tetramethylpiperidine, 98%
4-Hydroxy-2,2,6,6-tetramethylpiperidine, 98%
4-Hydroxy-2,2,6,6-tetramethylpiperidine, 98%
4-Hydroxy-2,2,6,6-tetramethylpiperidine, 98%
Thermo Scientific Chemicals

4-Hydroxy-2,2,6,6-tetramethylpiperidine, 98%

CAS: 2403-88-5 | C9H19NO | 157.257 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22664.22
also known as B22664-22
Price (USD)
355.65
Online Exclusive
395.00
Save 39.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
355.65
Online Exclusive
395.00
Save 39.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2403-88-5
IUPAC Name2,2,6,6-tetramethylpiperidin-4-ol
Molecular FormulaC9H19NO
InChI KeyVDVUCLWJZJHFAV-UHFFFAOYSA-N
SMILESCC1(C)CC(O)CC(C)(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Melting Point (clear melt)127.0-134.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only