3-Chlorothiophene, 98%, may contain up to 2% DMF
3-Chlorothiophene, 98%, may contain up to 2% DMF
3-Chlorothiophene, 98%, may contain up to 2% DMF
Thermo Scientific Chemicals

3-Chlorothiophene, 98%, may contain up to 2% DMF

CAS: 17249-80-8 | C4H3ClS | 118.578 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22669.06
also known as B22669-06
Price (USD)
44.65
Online Exclusive
49.60
Save 4.95 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.60
Save 4.95 (10%)
Each
Chemical Identifiers
CAS17249-80-8
IUPAC Name3-chlorothiophene
Molecular FormulaC4H3ClS
InChI KeyQUBJDMPBDURTJT-UHFFFAOYSA-N
SMILESClC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentDMF remains from the manufacturing process
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5% (excluding DMF)
Refractive Index1.5480-1.5540 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only