(+/-)-3-Amino-1,2-propanediol, 97%
(+/-)-3-Amino-1,2-propanediol, 97%
(+/-)-3-Amino-1,2-propanediol, 97%
Thermo Scientific Chemicals

(+/-)-3-Amino-1,2-propanediol, 97%

CAS: 616-30-8 | C3H9NO2 | 91.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B22672.14
also known as B22672-14
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.80
Save 11.15 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.80
Save 11.15 (15%)
Each
Chemical Identifiers
CAS616-30-8
IUPAC Name3-aminopropane-1,2-diol
Molecular FormulaC3H9NO2
InChI KeyKQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILESNCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormViscous liquid or low-melting solid
Assay from Supplier's CofA≥96.0%
3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhao, Y.; Zhang, S.; Zhang, Y.; Cui, S.; Chen, H.; Zhi, D.; Zhen, Y.; Zhang, S.; Huang, L. Tri-peptide cationic lipids for gene delivery. J. Mater. Chem. B 2015, 3 (1), 119-126.
  2. Lee, S.; Wang, W.; Lee, Y.; Sampson, N. S. Cyclic acetals as cleavable linkers for affinity capture. Org. Biomol. Chem. 2015, 13 (31), 8445-8452.