2-Amino-5-chlorobenzonitrile, 97%
2-Amino-5-chlorobenzonitrile, 97%
2-Amino-5-chlorobenzonitrile, 97%
Thermo Scientific Chemicals

2-Amino-5-chlorobenzonitrile, 97%

CAS: 5922-60-1 | C7H5ClN2 | 152.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22690.03
also known as B22690-03
Price (USD)
25.90
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
25.90
Each
Chemical Identifiers
CAS5922-60-1
IUPAC Name2-amino-5-chlorobenzonitrile
Molecular FormulaC7H5ClN2
InChI KeyQYRDWARBHMCOAG-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)92.5-98.5?C
Appearance (Color)Pale yellow to cream
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only