Phloroglucide, 95%
Phloroglucide, 95%
Phloroglucide, 95%
Thermo Scientific Chemicals

Phloroglucide, 95%

CAS: 491-45-2 | C12H10O5 | 234.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22693.14
also known as B22693-14
Price (USD)
852.65
Online Exclusive
947.00
Save 94.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
852.65
Online Exclusive
947.00
Save 94.35 (10%)
Each
Chemical Identifiers
CAS491-45-2
IUPAC Name[1,1'-biphenyl]-2,3',4,5',6-pentol
Molecular FormulaC12H10O5
InChI KeyKICYRZIVKKYRFS-UHFFFAOYSA-N
SMILESOC1=CC(O)=C(C(O)=C1)C1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pale yellow or pale pink
Assay (HPLC)≥94.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only