2-Bromoadamantane, 98%
2-Bromoadamantane, 98%
2-Bromoadamantane, 98%
Thermo Scientific Chemicals

2-Bromoadamantane, 98%

CAS: 7314-85-4 | C10H15Br | 215.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22760.14
also known as B22760-14
Price (USD)
171.65
Online Exclusive
191.00
Save 19.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
171.65
Online Exclusive
191.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS7314-85-4
IUPAC Name1-bromoadamantane
Molecular FormulaC10H15Br
InChI KeyVQHPRVYDKRESCL-UHFFFAOYSA-N
SMILESBrC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White
Melting Point (DSC)138.0-144.0?C
FormWaxy solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only