3,5-Diamino-1,2,4-triazole, 98%
3,5-Diamino-1,2,4-triazole, 98%
3,5-Diamino-1,2,4-triazole, 98%
3,5-Diamino-1,2,4-triazole, 98%
Thermo Scientific Chemicals

3,5-Diamino-1,2,4-triazole, 98%

CAS: 1455-77-2 | C2H5N5 | 99.097 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22775.06
also known as B22775-06
Price (USD)
39.65
Online Exclusive
43.80
Save 4.15 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.80
Save 4.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS1455-77-2
IUPAC Name1H-1,2,4-triazole-3,5-diamine
Molecular FormulaC2H5N5
InChI KeyPKWIYNIDEDLDCJ-UHFFFAOYSA-N
SMILESNC1=NC(N)=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to cream to pale brown or grey
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with acids and oxidizing agents.
RUO – Research Use Only

General References:

  1. Ren, Y-H.; Li, W.; Zhao, F-Q.; Yi, J-H.; Yan, B.; Ma, H-X.; Xu, K-Z.; Song, J-R.; Hu, R-Z. Crystal structure and thermal behaviors for 3,5-dinitrobenzoic acid of 3,5-diamino-1,2,4-triazole. J. Anal. Appl. Pyrolysis. 2013, 102, 89-96.
  2. Cadiau, A.; Martineau, C.; Taulelle, F.; Adil, K. Investigation of the composition space diagram of the ZnF2-3,5-diamino-1,2,4-triazole-HF-H2O chemical system and structural characterization of a new fluorinated guanazolate MOF [Zn3F2]·(Am2TAZ)4. J. Fluorine Chem. 2013, 150, 104-108.
  3. Bian, C.; Zhang, M.; Lia, C.; Zhou, Z. 3-Nitro-1-(2H-tetrazol-5-yl)-1H-1,2,4-triazol-5-amine (HANTT) and its energetic salts: highly thermally stable energetic materials with low sensitivity. J. Mater. Chem. A 2015, 3 (1), 163-169.
  4. Koshikawa, H.; Nakanishi,S.; Hashimoto, K.; Kamiya, K. Heat-treated 3,5-diamino-1,2,4-triazole/graphene hybrid functions as an oxygen reduction electrocatalyst with high activity and stability. Electrochim. Acta. 2015, 180, 173-177.
  5. Ge, J.; Yang, Q.; Xie, G.; Chen, S.; Gao, S. Synthesis, structure and thermochemical study of a cobalt energetic coordination compound incorporating 3,5-diamino-1,2,4-triazole and pyridine-2,6-dicarboxylic acid. J. Chem. Thermodyn. 2015, 80, 1-6.