2,3-Dichlorobenzeneboronic acid, 98%
2,3-Dichlorobenzeneboronic acid, 98%
2,3-Dichlorobenzeneboronic acid, 98%
Thermo Scientific Chemicals

2,3-Dichlorobenzeneboronic acid, 98%

CAS: 151169-74-3 | C6H5BCl2O2 | 190.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22781.14
also known as B22781-14
Price (USD)
243.65
Online Exclusive
271.00
Save 27.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
243.65
Online Exclusive
271.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS151169-74-3
IUPAC Name(2,3-dichlorophenyl)boronic acid
Molecular FormulaC6H5BCl2O2
InChI KeyTYIKXPOMOYDGCS-UHFFFAOYSA-N
SMILESOB(O)C1=C(Cl)C(Cl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only