2-Methoxybenzoyl chloride, 97%
2-Methoxybenzoyl chloride, 97%
2-Methoxybenzoyl chloride, 97%
Thermo Scientific Chemicals

2-Methoxybenzoyl chloride, 97%

CAS: 21615-34-9 | C8H7ClO2 | 170.592 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22784.06
also known as B22784-06
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.80
Save 7.15 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.80
Save 7.15 (14%)
Each
Chemical Identifiers
CAS21615-34-9
IUPAC Name2-methoxybenzoyl chloride
Molecular FormulaC8H7ClO2
InChI KeyRZNHSEZOLFEFGB-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5695-1.5745 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only