2,6-Difluorobenzeneboronic acid, 98%
2,6-Difluorobenzeneboronic acid, 98%
2,6-Difluorobenzeneboronic acid, 98%
2,6-Difluorobenzeneboronic acid, 98%
Thermo Scientific Chemicals

2,6-Difluorobenzeneboronic acid, 98%

CAS: 162101-25-9 | C6H5BF2O2 | 157.911 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22805.06
also known as B22805-06
Price (USD)
70.65
Online Exclusive
78.30
Save 7.65 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.30
Save 7.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS162101-25-9
IUPAC Name(2,6-difluorophenyl)boronic acid
Molecular FormulaC6H5BF2O2
InChI KeyDBZAICSEFBVFHL-UHFFFAOYSA-N
SMILESOB(O)C1=C(F)C=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only