Bromosuccinic acid, 96%
Bromosuccinic acid, 96%
Bromosuccinic acid, 96%
Thermo Scientific Chemicals

Bromosuccinic acid, 96%

CAS: 923-06-8 | C4H5BrO4 | 196.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22829.06
also known as B22829-06
Price (USD)
45.65
Online Exclusive
50.50
Save 4.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.50
Save 4.85 (10%)
Each
Chemical Identifiers
CAS923-06-8
IUPAC Name2-bromobutanedioic acid
Molecular FormulaC4H5BrO4
InChI KeyQQWGVQWAEANRTK-UHFFFAOYNA-N
SMILESOC(=O)CC(Br)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥95.0%
Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.

Solubility
Insoluble in ether.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A. R. Olson; L. D. Frashier; Frances J. Spieth. The Racemization of the Dimethyl Ester of l-Bromosuccinic Acid by Lithium Bromide in Acetone. J. Phys. Chem. 1951, 55 (6), 860-873.
  2. Tadashi Shiraiwa; Masanori Ohkubo; Hideya Miyazaki; Motoki Kubo; Hiroki Nishigawa; Toshihiro Tsujimoto; Hidemoto Kurokawa. Optical Resolution by Preferential Crystallization of (RS)-Bromosuccinic Acid. Bulletin of the Chemical Society of Japan. 1998, 71, (3), 735-739.