4-Chlorobenzyl bromide, 98+%
4-Chlorobenzyl bromide, 98+%
4-Chlorobenzyl bromide, 98+%
Thermo Scientific Chemicals

4-Chlorobenzyl bromide, 98+%

CAS: 622-95-7 | C7H6BrCl | 205.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22858.06
also known as B22858-06
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.30
Save 7.65 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.30
Save 7.65 (14%)
Each
Chemical Identifiers
CAS622-95-7
IUPAC Name1-(bromomethyl)-4-chlorobenzene
Molecular FormulaC7H6BrCl
InChI KeyKQNBRMUBPRGXSL-UHFFFAOYSA-N
SMILESClC1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline powder or fused solid
Appearance (Color)White
Assay (GC)>98.0%
4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
RUO – Research Use Only

General References:

  1. Masaru Ogata; Hiroshi Matsumoto; Sumio Shimizu; Shiro Kida; Motoo Shiro; Katsuya Tawara. Synthesis and antifungal activity of new 1-vinylimidazoles. J. Med. Chem. 1987, 30,(8), 1348-1354.
  2. Christian Beaulieu; Zhaoyin Wang; Danielle Denis; Gillian Greig; Sonia Lamontagne; Gary O'Neill; Deborah Slipetz; Jennifer Wang. Benzimidazoles as new potent and selective DP antagonists for the treatment of allergic rhinitis. Bioorganic & Medicinal Chemistry Letters. 2004, 14,(12), 3195-3199.