1-Naphthyl phosphate disodium salt hydrate, 99%
1-Naphthyl phosphate disodium salt hydrate, 99%
1-Naphthyl phosphate disodium salt hydrate, 99%
Thermo Scientific Chemicals

1-Naphthyl phosphate disodium salt hydrate, 99%

1-Naphthyl phosphate disodium salt hydrate, 99%, C10H9Na2O5P, CAS Number-207569-06-0 | CAS: 207569-06-0 | C10H7Na2O4P | 268.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22866.06
also known as B22866-06
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS207569-06-0
IUPAC Namedisodium naphthalen-1-yl phosphate
Molecular FormulaC10H7Na2O4P
InChI KeyQYURIFWAOPAPAJ-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)3.10-9.40% (0.5-1.5 waters)
Identification (FTIR)Conforms
Appearance (Color)White to cream or pale pink or pale brown
FormPowder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (dry wt. basis)
1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
Substrate for phosphatases: Methods of Enzymatic Analysis, 3rd ed., H. U. Bergmeyer, Ed., Verlag Chemie, Weinheim (1984), vol. 4, p102.