1,2-Bis(2-chloroethoxy)ethane, 97%
1,2-Bis(2-chloroethoxy)ethane, 97%
1,2-Bis(2-chloroethoxy)ethane, 97%
Thermo Scientific Chemicals

1,2-Bis(2-chloroethoxy)ethane, 97%

CAS: 112-26-5 | C6H12Cl2O2 | 187.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B22884.36
also known as B22884-36
Price (USD)
134.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
134.00
Each
Chemical Identifiers
CAS112-26-5
IUPAC Name1-chloro-2-[2-(2-chloroethoxy)ethoxy]ethane
Molecular FormulaC6H12Cl2O2
InChI KeyAGYUOJIYYGGHKV-UHFFFAOYSA-N
SMILESClCCOCCOCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4590-1.4640 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
1,2-Bis(2-chloroethoxy)ethane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(2-chloroethoxy)ethane is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. F.Teixidor; C.Vinas; J.Rius; C.Miravitlles; J.Casabo. A new type of macrocycle incorporating closo- and nido-carborane cages: molecular structures of 1,2-(1,10-dithia-4,7-dioxadecane-1,10-diyl)-1,2-dicarba-closo-dodecaborane and sodium 7,8-(1,13-dithia-4,7,10-trioxatridecane-1,13-diyl)-7,8-dicarbanido-undecaborate(12). Inorg. Chem. 1990, 29,(1), 149-152.
  2. R.M.Izatt; R.E.Terry; D.P.Nelson; Y.Chan; D.J.Eatough; J.S.Bradshaw; L.D.Hansen; J.J.Christensen. Calorimetric titration study of the interaction of some uni- and bivalent cations with benzo-15-crown-5, 18-crown-6, dibenzo-24-crown-8, and dibenzo-27-crown-9 in methanol-water solvents, at 25.degree.C and .mu. = 0.1. J. Am. Chem. Soc. 1976, 98,(24), 7626-7630.