4-(Trifluoromethylsulfonyl)benzonitrile, 97+%
4-(Trifluoromethylsulfonyl)benzonitrile, 97+%
4-(Trifluoromethylsulfonyl)benzonitrile, 97+%
4-(Trifluoromethylsulfonyl)benzonitrile, 97+%
Thermo Scientific Chemicals

4-(Trifluoromethylsulfonyl)benzonitrile, 97+%

CAS: 312-21-0 | C8H4F3NO2S | 235.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22907.03
also known as B22907-03
Price (USD)
42.65
Online Exclusive
47.50
Save 4.85 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
42.65
Online Exclusive
47.50
Save 4.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS312-21-0
IUPAC Name4-trifluoromethanesulfonylbenzonitrile
Molecular FormulaC8H4F3NO2S
InChI KeyUHCGSMMNICMDIX-UHFFFAOYSA-N
SMILESFC(F)(F)S(=O)(=O)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Melting Point (clear melt)88.0-97.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)Cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only