(+/-)-Tetrahydro-2-furoic acid, 97+%
(+/-)-Tetrahydro-2-furoic acid, 97+%
(+/-)-Tetrahydro-2-furoic acid, 97+%
Thermo Scientific Chemicals

(+/-)-Tetrahydro-2-furoic acid, 97+%

CAS: 16874-33-2 | C5H8O3 | 116.116 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B22927.14
also known as B22927-14
Price (USD)
59.65
Online Exclusive
66.20
Save 6.55 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
59.65
Online Exclusive
66.20
Save 6.55 (10%)
Each
Chemical Identifiers
CAS16874-33-2
IUPAC Nameoxolane-2-carboxylic acid
Molecular FormulaC5H8O3
InChI KeyUJJLJRQIPMGXEZ-UHFFFAOYNA-N
SMILESOC(=O)C1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4575-1.4625 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Assay (Silylated GC)≥97.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only