2,3-Dichlorophenol, 98+%
2,3-Dichlorophenol, 98+%
2,3-Dichlorophenol, 98+%
Thermo Scientific Chemicals

2,3-Dichlorophenol, 98+%

CAS: 576-24-9 | C6H4Cl2O | 163.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B22943.36
also known as B22943-36
Price (USD)
273.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
273.00
Each
Chemical Identifiers
CAS576-24-9
IUPAC Name2,3-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyUMPSXRYVXUPCOS-UHFFFAOYSA-N
SMILESOC1=CC=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)White to cream
FormLumps or fused solid or chunks
Melting Point (clear melt)55.0-61.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only