L-(-)-Bornyl acetate, 95%
L-(-)-Bornyl acetate, 95%
L-(-)-Bornyl acetate, 95%
Thermo Scientific Chemicals

L-(-)-Bornyl acetate, 95%

CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B22957.14
also known as B22957-14
Price (USD)
23.65
Online exclusive
25.60
Save 1.95
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
23.65
Online exclusive
25.60
Save 1.95
Each
Chemical Identifiers
CAS5655-61-8
IUPAC Name(1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
Molecular FormulaC12H20O2
InChI KeyKGEKLUUHTZCSIP-HOSYDEDBSA-N
SMILESCC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94%
Identification (FTIR)Conforms
Impurity contentisobornyl acetate ≤ 4%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only