alpha-Acetyl-gamma-butyrolactone, 98+%
alpha-Acetyl-gamma-butyrolactone, 98+%
alpha-Acetyl-gamma-butyrolactone, 98+%
Thermo Scientific Chemicals

alpha-Acetyl-gamma-butyrolactone, 98+%

CAS: 517-23-7 | C6H8O3 | 128.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B22967.22
also known as B22967-22
Price (USD)
28.65
Online Exclusive
31.90
Save 3.25 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
28.65
Online Exclusive
31.90
Save 3.25 (10%)
Each
Chemical Identifiers
CAS517-23-7
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥98.0%
Appearance (Color)Clear colorless
Refractive Index1.4575-1.4615 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only