3-Methoxy-4-methylbenzoic acid, 99%
3-Methoxy-4-methylbenzoic acid, 99%
3-Methoxy-4-methylbenzoic acid, 99%
Thermo Scientific Chemicals

3-Methoxy-4-methylbenzoic acid, 99%

CAS: 7151-68-0 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22982.06
also known as B22982-06
Price (USD)
74.65
Special offer
Online exclusive
Ends: 28-Dec-2026
88.20
Save 13.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 28-Dec-2026
88.20
Save 13.55
Each
Chemical Identifiers
CAS7151-68-0
IUPAC Name3-methoxy-4-methylbenzoic acid
Molecular FormulaC9H10O3
InChI KeyCEAVPXDEPGAVDA-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystalline powder
Assay (Aqueous acid-base Titration)>98.5 to <101.5%
Assay (Silylated GC)>98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only