Thymol Blue sodium salt
Thymol Blue sodium salt
Thymol Blue sodium salt
Thermo Scientific Chemicals

Thymol Blue sodium salt

CAS: 62625-21-2 | C27H29NaO5S | 488.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B23007.30
also known as B23007-30
Price (USD)
518.65
Online Exclusive
576.00
Save 57.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
518.65
Online Exclusive
576.00
Save 57.35 (10%)
Each
Chemical Identifiers
CAS62625-21-2
IUPAC Namesodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
Molecular FormulaC27H29NaO5S
InChI KeyOCMIKNBSIRSUPI-FRWNMSGJSA-M
SMILES[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color ChangeVisual transition: pH 8.0 (Yellow) - pH 9.6 (Blue)
lambda-max (UV-Vis)593 - 601nm (0.01M NaOH)
Appearance (Color)Orange to Dark Orange to Red-Brown to Dark green-Dark Brown
FormPowder
Specific Absorbance (E 1%/1cm)≥ 550 at Lambda max
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only