3-Methylbenzeneboronic acid, 97%
3-Methylbenzeneboronic acid, 97%
3-Methylbenzeneboronic acid, 97%
Thermo Scientific Chemicals

3-Methylbenzeneboronic acid, 97%

CAS: 17933-03-8 | C7H9BO2 | 135.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23025.03
also known as B23025-03
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.60
Save 6.95
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.60
Save 6.95
Each
Chemical Identifiers
CAS17933-03-8
IUPAC Name(3-methylphenyl)boronic acid
Molecular FormulaC7H9BO2
InChI KeyBJQCPCFFYBKRLM-UHFFFAOYSA-N
SMILESCC1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)152.0-162.0?C
View more
Suzuki coupling reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only