p-Tolunitrile, 98+%
p-Tolunitrile, 98+%
p-Tolunitrile, 98+%
Thermo Scientific Chemicals

p-Tolunitrile, 98+%

CAS: 104-85-8 | C8H7N | 117.151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number B23029.30
also known as B23029-30
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS104-85-8
IUPAC Name4-methylbenzonitrile
Molecular FormulaC8H7N
InChI KeyVCZNNAKNUVJVGX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystalline mass or fused solid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only