(4-Bromobutyl)triphenylphosphonium bromide, 98%
(4-Bromobutyl)triphenylphosphonium bromide, 98%
(4-Bromobutyl)triphenylphosphonium bromide, 98%
(4-Bromobutyl)triphenylphosphonium bromide, 98%
Thermo Scientific Chemicals

(4-Bromobutyl)triphenylphosphonium bromide, 98%

CAS: 7333-63-3 | C22H23Br2P | 478.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B23040.09
also known as B23040-09
Price (USD)
41.70
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
41.70
Each
Add to cart
Chemical Identifiers
CAS7333-63-3
IUPAC Name(4-bromobutyl)triphenylphosphanium bromide
Molecular FormulaC22H23Br2P
InChI KeyBJDNCJGRAMGIRU-UHFFFAOYSA-M
SMILES[Br-].BrCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1.0%
Melting Point (clear melt)202.5-208.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only