4-Fluoroanisole, 99%, Thermo Scientific Chemicals
4-Fluoroanisole, 99%, Thermo Scientific Chemicals
4-Fluoroanisole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoroanisole, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B23048.22
also known as B23048-22
100 g
Catalog number B23048.22
also known as B23048-22
Price (USD)
122.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
122.00
Each
Chemical Identifiers
CAS459-60-9
IUPAC Name1-fluoro-4-methoxybenzene
Molecular FormulaC7H7FO
InChI KeyVIPWUFMFHBIKQI-UHFFFAOYSA-N
SMILESCOC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Refractive Index1.4865-1.4895
Assay (GC)>98.5%
Formliquid
4-Fluoroanisole is a raw material for medicine and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoroanisole is a raw material for medicine and intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. D.G. Lister. The barrier to internal rotation of the methoxy group in anisoles: evidence from the microwave spectrum of p-fluoroanisole. Journal of Molecular Structure. 1980, 68, 33-40.
  2. D.W. Slocum; D.S. Coffey; A. Siegel; P. Grimes. A predicative model for certain directed metalations, II; application to the behavior of p-fluoroanisole. Tetrahedron Letters. 1994, 35 (3), 389-392.