3-Dimethylaminophenol, 97+%
3-Dimethylaminophenol, 97+%
3-Dimethylaminophenol, 97+%
Thermo Scientific Chemicals

3-Dimethylaminophenol, 97+%

CAS: 99-07-0 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23067.06
also known as B23067-06
Price (USD)
29.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.20
Save 4.55 (13%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.20
Save 4.55 (13%)
Each
Chemical Identifiers
CAS99-07-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown to dark brown to purple to dark purple
FormPowder
Assay (GC)≥97.0%
Identification (FTIR)Conforms
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents, light.
RUO – Research Use Only

General References:

  1. Bozena Bukowska; Jaromir Michałowicz; Wirgiliusz Duda. Alterations in human red blood cell properties induced by 3-(dimethylamino)phenol (in vitro). Toxicology in Vitro. 2007, 21, (8), 1574-1580.
  2. Kling RE. UV spectrophotometric assay of neostigmine methylsulfate as the alkaline hydrolysis product, 3-dimethylaminophenol. J Assoc Off Anal Chem. 1971, 54 (1), 21-6.