(1R,3S)-(+)-Camphoric acid, 98+%
(1R,3S)-(+)-Camphoric acid, 98+%
(1R,3S)-(+)-Camphoric acid, 98+%
(1R,3S)-(+)-Camphoric acid, 98+%
Thermo Scientific Chemicals

(1R,3S)-(+)-Camphoric acid, 98+%

CAS: 124-83-4 | C10H14O4-2 | 198.218 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23106.36
also known as B23106-36
Price (USD)
285.65
Online Exclusive
317.00
Save 31.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
285.65
Online Exclusive
317.00
Save 31.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS124-83-4
IUPAC Name(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
Molecular FormulaC10H14O4
InChI KeyLSPHULWDVZXLIL-LDWIPMOCSA-L
SMILESCC1(C)[C@H](CC[C@@]1(C)C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)184.5-190.5?C
Optical Rotation46 ± 2? (c=10 in ethanol)
Appearance (Color)White
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only