Methyltrichlorosilane, 97%
Methyltrichlorosilane, 97%
Methyltrichlorosilane, 97%
Thermo Scientific Chemicals

Methyltrichlorosilane, 97%

CAS: 75-79-6 | CH3Cl3Si | 149.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B23107.22
also known as B23107-22
Price (USD)
39.20
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
39.20
Each
Chemical Identifiers
CAS75-79-6
IUPAC Nametrichloro(methyl)silane
Molecular FormulaCH3Cl3Si
InChI KeyJLUFWMXJHAVVNN-UHFFFAOYSA-N
SMILESC[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4090-1.4140 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Methyltrichlorosilane is used in production of methyl silicone resins, its vapor reacts with water on surfaces to give a thin layer of methylpolysiloxane which make it a water-repellent film. A combination of methyltrichlorosilane and sodium iodide can be used to cleave carbon-oxygen bonds such as methyl ethers. It is used as a precursor for forming various cross-linked siloxane polymers. Trichloromethylsilane is the starting material for the production of pure silicon for manufacture of semiconductors and optical fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltrichlorosilane is used in production of methyl silicone resins, its vapor reacts with water on surfaces to give a thin layer of methylpolysiloxane which make it a water-repellent film. A combination of methyltrichlorosilane and sodium iodide can be used to cleave carbon-oxygen bonds such as methyl ethers. It is used as a precursor for forming various cross-linked siloxane polymers. Trichloromethylsilane is the starting material for the production of pure silicon for manufacture of semiconductors and optical fibers.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

General References:

  1. Ching Yi Tsaia; Seshu B. Desua and Chien C. Chiu. Kinetic study of silicon carbide deposited from methyltrichlorosilane precursor. Journal of Materials Research. 1994, 9, (1), 104-111.
  2. Th. Kunstmann and S. Vepřek. Heteroepitaxy of β-SiC from methyltrichlorosilane and methyltribromosilane on Si(100) without a carbon buffer layer. Appl. Phys. Lett. 1995, 67 (21), 3126.